Reagents Sodium Borohydride, Wide-Mouth Amber Glass Bottle, 25 g

Representative image only.

Reagents Sodium Borohydride, Wide-Mouth Amber Glass Bottle, 25 g

Reagents | Mfr: C2310550-25C1| Item: UX-62285-63

Manufactured using the highest quality raw materials in ISO 9001 facility

$95.80USD / Each

$95.80USD / Each
Product Description

Key Features

  • Minimum Assay 98%
  • White to gray solid
  • Density: 1.07
  • Boiling point: approximately 400 °C (752 °F), decomposes slowly
  • Melting point: >300 °C (>572 °F), decomposes
  • Solubility information: decomposes in water
  • 25 g wide-mouth amber glass bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Linear FormulaNaBH4
  • ClassSalts
  • ChemistryInorganics
  • SynonymsSodium tetrahydridoborate
  • FormSolid
  • Weight (g)25
  • Formula Weight37.83
  • PackagingWide-mouth amber glass bottle
  • DOT InformationSODIUM BOROHYDRIDE, 4.3, P.G. I
  • Storage SymbolAmbient
  • CAS Number16940-66-2
  • UN NumberUN1426
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • DescriptionSodium Borohydride, Wide-Mouth Amber Glass Bottle, 25 g
Documents & Certificates
Reviews
Q&A