Reagents Sodium Carbonate Anhydrous, Purified, Wide-Mouth Container, 500 g

Representative image only.

Reagents Sodium Carbonate Anhydrous, Purified, Wide-Mouth Container, 500 g

Reagents | Mfr: C2312100-500A5| Item: UX-78913-64

Manufactured using the highest quality raw materials in ISO 9001 facility

$39.20USD / Each

$39.20USD / Each
Product Description

Key Features

  • Colorless/white granules
  • Density: 2.53
  • Boiling point: decomposes
  • Solubility Information: 215 g/L at 20 °C (68 °F)
  • pH range: 11 to 12
  • 500 g wide-mouth container
  • 10 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

10 Year Warranty
Specifications
  • Purity GradePurified
  • Linear FormulaNa2CO3
  • ClassSalts
  • FormSolid
  • SynonymsSoda ash
  • Weight (g)500
  • Formula Weight105.99
  • PackagingWide-mouth container
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number497-19-8
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • Melting Point C (° C)851
  • Melting Point F (° F)1563.8
  • DescriptionSodium Carbonate Anhydrous, Purified, Wide-Mouth Container, 500 g
  • Warranty10 year
Documents & Certificates
Reviews
Q&A