Reagents Sodium Chromate Anhydrous, Purified, Wide-Mouth Amber Glass Bottle, 100 g

Representative image only.

Reagents Sodium Chromate Anhydrous, Purified, Wide-Mouth Amber Glass Bottle, 100 g

Reagents | Mfr: C2314000-100C2| Item: UX-62285-71

Manufactured using the highest quality raw materials in ISO 9001 facility

$36.50USD / Each

$36.50USD / Each
Product Description

Key Features

  • Yellow solid
  • Density: 2.72
  • Solubility information: 876 g/L at 25 °C (77 °F)
  • 100 g wide-mouth amber glass bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradePurified
  • Linear FormulaNa2CrO4
  • ClassSalts
  • ChemistryInorganics
  • FormSolid
  • Weight (g)100
  • Formula Weight161.9732
  • PackagingWide-mouth amber glass bottle
  • DOT InformationOXIDIZING SOLID, CORROSIVE, N.O.S. (SODIUM CHROMATE), 5.1 (8), P.G. III
  • Storage SymbolAmbient
  • CAS Number7775-11-3
  • UN NumberUN3085
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • Melting Point C (° C)792
  • Melting Point F (° F)1457.6
  • DescriptionSodium Chromate Anhydrous, Purified, Wide-Mouth Amber Glass Bottle, 100 g
Documents & Certificates
Reviews
Q&A