Reagents Sodium Fluoride, Purified, Wide-Mouth Amber Glass Bottle, 125 g

Representative image only.

Reagents Sodium Fluoride, Purified, Wide-Mouth Amber Glass Bottle, 125 g

Reagents | Mfr: C2317000-125D2| Item: UX-62285-76

Manufactured using the highest quality raw materials in ISO 9001 facility

$45.50USD / Each

$45.50USD / Each
Product Description

Key Features

  • White solid
  • Density: 2.78
  • Boiling point: approximately 1704 °C (3099 °F)
  • Solubility nformation: 42.2 g/L at 20 °C (68 °F)
  • 125 g wide-mouth amber glass bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradePurified
  • Linear FormulaNaF
  • ClassSalts
  • ChemistryInorganics
  • FormSolid
  • Weight (g)125
  • Formula Weight41.99
  • PackagingWide-mouth amber glass bottle
  • DOT InformationSODIUM FLUORIDE, SOLID, 6.1, P.G. III
  • Storage SymbolAmbient
  • UN NumberUN1690
  • CAS Number7681-49-4
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • Melting Point C (° C)990
  • Melting Point F (° F)1814
  • DescriptionSodium Fluoride, Purified, Wide-Mouth Amber Glass Bottle, 125 g
Documents & Certificates
Reviews
Q&A