Reagents Sodium Phosphate Dibasic Anhydrous, ACS Reagent, Wide-Mouth Amber Glass Bottle, 100 g

Representative image only.

Reagents Sodium Phosphate Dibasic Anhydrous, ACS Reagent, Wide-Mouth Amber Glass Bottle, 100 g

Reagents | Mfr: C1324050-100C2| Item: UX-78914-02

Manufactured using the highest quality raw materials in ISO 9001 facility

$45.80USD / Each

$45.80USD / Each
Product Description

Key Features

  • Minimum Assay 99%
  • Colorless/white powder
  • Density: 2.07
  • Melting poing: >240 °C (464 °F), decomposes
  • Solubility information: 118 g/L at 25 °C (77 °F)
  • pH range: 8.7 to 9.3
  • 100 g wide-mouth amber glass bottle
  • 5 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

5 Year Warranty
Specifications
  • Purity GradeACS
  • Linear FormulaNa2HPO4
  • ClassSalts
  • SynonymsDisodium hydrogen phosphate
  • FormSolid
  • Weight (g)100
  • Formula Weight141.96
  • PackagingWide-mouth amber glass bottle
  • CAS Number7558-79-4
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • DescriptionSodium Phosphate Dibasic Anhydrous, ACS Reagent, Wide-Mouth Amber Glass Bottle, 100 g
  • Warranty5 years
Documents & Certificates
Reviews
Q&A