Reagents Sodium Phosphate Dibasic Anhydrous, Purified, Natural Poly Container, 2.5 kg

Representative image only.

Reagents Sodium Phosphate Dibasic Anhydrous, Purified, Natural Poly Container, 2.5 kg

Reagents | Mfr: C2324000-2.5A5| Item: UX-78919-43

Manufactured using the highest quality raw materials in ISO 9001 facility

$237.25USD / Each

$237.25USD / Each
Product Description

Key Features

  • Colorless/white granules
  • Density: 2.07
  • Melting point: >240 °C (464 °F), decomposes
  • Solubility information: 118 g/L at 25 °C (77 °F)
  • pH range: 8.7 to 9.3
  • 2.5 kg natural poly container
  • 10 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

10 Year Warranty
Specifications
  • Purity GradePurified
  • Linear FormulaNa2HPO4
  • ClassSalts
  • SynonymsDisodium hydrogen phosphate
  • FormSolid
  • Weight (g)2500
  • Formula Weight141.96
  • PackagingPoly container
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number7558-79-4
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • DescriptionSodium Phosphate Dibasic Anhydrous, Purified, Natural Poly Container, 2.5 kg
  • Warranty10 year
Documents & Certificates
Reviews
Q&A