Reagents Sodium Phosphate Monobasic Anhydrous, Purified, Wide-Mouth Container, 500 g

Representative image only.

Reagents Sodium Phosphate Monobasic Anhydrous, Purified, Wide-Mouth Container, 500 g

Reagents | Mfr: C2323500-500A5| Item: UX-78914-04

Manufactured using the highest quality raw materials in ISO 9001 facility

$89.40USD / Each

$89.40USD / Each
Product Description

Key Features

  • White granules
  • Density: 2.04
  • Solubility information: 949 g/L at 25 °C (77 °F)
  • 500 g wide-mouth container
  • 10 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

10 Year Warranty
Specifications
  • Purity GradePurified
  • Linear FormulaNaH2PO4
  • ClassSalts
  • SynonymsSodium dihydrogen phosphate
  • FormSolid
  • Weight (g)500
  • Formula Weight119.98
  • PackagingWide-mouth container
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number7558-80-7
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • Melting Point C (° C)60
  • Melting Point F (° F)140
  • DescriptionSodium Phosphate Monobasic Anhydrous, Purified, Wide-Mouth Container, 500 g
  • Warranty10 year
Documents & Certificates
Reviews
Q&A