Reagents Sodium Phosphate Monobasic Monohydrate, ACS Reagent, Wide-Mouth Amber Glass Bottle, 100 g

Representative image only.

Reagents Sodium Phosphate Monobasic Monohydrate, ACS Reagent, Wide-Mouth Amber Glass Bottle, 100 g

Reagents | Mfr: C1323500-100C2| Item: UX-78914-05

Manufactured using the highest quality raw materials in ISO 9001 facility

$56.60USD / Each

$56.60USD / Each
Product Description

Key Features

  • Minimum Assay 98%
  • Colorless/white crystals
  • Density: 2.04
  • Boiling point: approximately 204 °C (399.2 °F)
  • Solubility information: 949 g/L at 25 °C (77 °F)
  • 100 g wide-mouth amber glass bottle
  • 7 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaNaH2PO4·H2O
  • ClassSalts
  • SynonymsSodium phosphate monobasic 1-hydrate, Sodium dihydrogen phosphate monohydrate
  • FormSolid
  • Weight (g)100
  • Formula Weight137.99
  • PackagingWide-mouth amber glass bottle
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number10049-21-5
  • Shelf Life (days)2585
  • Shelf Life (months)84
  • Melting Point C (° C)100
  • Melting Point F (° F)212
  • DescriptionSodium Phosphate Monobasic Monohydrate, ACS Reagent, Wide-Mouth Amber Glass Bottle, 100 g
  • Warranty7 year
Documents & Certificates
Reviews
Q&A