Reagents Sodium Silicate, 42° Baumé, Natural Poly Container, 4 L

Representative image only.

Reagents Sodium Silicate, 42° Baumé, Natural Poly Container, 4 L

Reagents | Mfr: C2325000-4A| Item: UX-78925-07

Manufactured using the highest quality raw materials in ISO 9001 facility

$89.40USD / Each

$89.40USD / Each
Product Description

Key Features

  • 42° Baumé
  • Colorless/white liquid
  • Density: 1.39
  • Solubility information: miscible
  • 4 L natural poly container
  • 5 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

5 Year Warranty
Specifications
  • ClassSolution
  • ChemistryAnalytical Reagents
  • FormLiquid
  • Volume (mL)4000
  • PackagingPoly container
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number1344-09-8
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • Boiling Point C (° C)102
  • Boiling Point F (° F)215.6
  • DescriptionSodium Silicate, 42° Baumé, Natural Poly Container, 4 L
  • Warranty5 years
Documents & Certificates
Reviews
Q&A