Reagents Sodium Sulfate Anhydrous, Purified, Natural Poly Container, 3 kg

Representative image only.

Reagents Sodium Sulfate Anhydrous, Purified, Natural Poly Container, 3 kg

Reagents | Mfr: C2325500-3A5| Item: UX-62285-99

Manufactured using the highest quality raw materials in ISO 9001 facility

$72.20USD / Each

$72.20USD / Each
Product Description

Key Features

  • White solid
  • Density: 2.7
  • Boiling point: >890 °C (>1634 °F)
  • Melting point: 884 to 888 °C (1623.2 to 1630.4 °F)
  • Solubility information: 200 g/L at 20 °C (68 °F)
  • pH range: 5.2 to 9.2
  • 3 kg natural poly container

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradePurified
  • Linear FormulaNa2SO4
  • ClassSalts
  • ChemistryInorganics
  • FormSolid
  • Weight (g)3000
  • Formula Weight142.04
  • PackagingPoly container
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number7757-82-6
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • DescriptionSodium Sulfate Anhydrous, Purified, Natural Poly Container, 3 kg
Documents & Certificates
Reviews
Q&A