Reagents Sodium Sulfate Anhydrous, Purified, Wide-Mouth Container, 500 g

Representative image only.

Reagents Sodium Sulfate Anhydrous, Purified, Wide-Mouth Container, 500 g

Reagents | Mfr: C2325500-500A5| Item: UX-78914-10

Manufactured using the highest quality raw materials in ISO 9001 facility

$34.40USD / Each

$34.40USD / Each
Product Description

Key Features

  • White solid
  • Density: 2.7
  • Boiling point: >890 °C (>1634 °F)
  • Melting point: 884 to 888 °C (1623.2 to 1630.4 °F)
  • Solubility information: 200 g/L at 20 °C (68 °F)
  • pH range: 5.2 to 9.2
  • 500 g wide-mouth container
  • 10 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

10 Year Warranty
Specifications
  • Purity GradePurified
  • Linear FormulaNa2SO4
  • ClassSalts
  • FormSolid
  • Weight (g)500
  • Formula Weight142.04
  • PackagingWide-mouth container
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number7757-82-6
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • DescriptionSodium Sulfate Anhydrous, Purified, Wide-Mouth Container, 500 g
  • Warranty10 year
Documents & Certificates
Reviews
Q&A