Reagents Sodium Tartrate Dihydrate, ACS Reagent, 2Kg

Representative image only.

Reagents Sodium Tartrate Dihydrate, ACS Reagent, 2Kg

Reagents | Mfr: C1327000-2A5| Item: UX-62283-19

Manufactured using the highest quality raw materials in ISO 9001 facility

$447.00USD / Each

$447.00USD / Each
Product Description

Key Features

  • Minimum Assay 99%
  • Colorless/white crystals
  • Density: 1.82
  • Solubility information: 330 g/L

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaC4H8Na2O8
  • ClassSalts
  • ChemistryInorganics
  • SynonymsSodium tartrate 2-hydrate
  • FormSolid
  • Weight (g)2000
  • Formula Weight230.08
  • PackagingPoly container
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number6106-24-7
  • Shelf Life (days)2585
  • Shelf Life (months)84
  • DescriptionSodium Tartrate Dihydrate, ACS Reagent, Natural Poly Container, 2 kg
Documents & Certificates
Reviews
Q&A