Reagents Starch, Potato, Wide-Mouth Container, 500 g

Representative image only.

Reagents Starch, Potato, Wide-Mouth Container, 500 g

Reagents | Mfr: C2331500-500A5| Item: UX-78914-26

Manufactured using the highest quality raw materials in ISO 9001 facility

$305.50USD / Each

$305.50USD / Each
Product Description

Key Features

  • White solid
  • Density: 1.5
  • Melting point: 255 to 258 °C (491 to 496.4 °F)
  • Solubility information: insoluble
  • 500 g wide-mouth container
  • 10 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

10 Year Warranty
Specifications
  • Linear Formula(C6H10O5)n
  • ClassSalts
  • FormSolid
  • Weight (g)500
  • PackagingWide-mouth container
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number9005-84-9
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • DescriptionStarch, Potato, Wide-Mouth Container, 500 g
  • Warranty10 year
Documents & Certificates
Reviews
Q&A