Reagents Sulfuric Acid, 66° Baumé, Technical, Natural Poly Bottle, 4 L

Representative image only.

Reagents Sulfuric Acid, 66° Baumé, Technical, Natural Poly Bottle, 4 L

Reagents | Mfr: C5102700-4A| Item: UX-78925-52

Manufactured using the highest quality raw materials in ISO 9001 facility

$97.10USD / Each

$97.10USD / Each
Product Description

Key Features

  • Minimum Assay 93%
  • Colorless liquid
  • Density: 1.83
  • Solubility information: miscible
  • pH range: <1
  • 4 L natural poly bottle
  • 20 years warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

20 Year Warranty
Specifications
  • Purity GradeTechnical
  • Linear FormulaH2SO4
  • ClassAcid
  • FormLiquid
  • Volume (mL)4000
  • Formula Weight98.0785
  • Vapor Pressure (torr)< 0.1
  • PackagingPoly bottle
  • DOT InformationSULFURIC ACID, 8, P.G. II
  • Storage SymbolAmbient
  • CAS Number7664-93-9
  • UN NumberUN1830
  • Shelf Life (days)760
  • Shelf Life (months)240
  • Boiling Point C (° C)335
  • Boiling Point F (° F)635
  • Melting Point C (° C)-20
  • Melting Point F (° F)-4
  • DescriptionSulfuric Acid, 66° Baumé, Technical, Natural Poly Bottle, 4 L
  • Warranty20 years
Documents & Certificates
Reviews
Q&A