Reagents Thymol Blue Indicator, 0.04% (w/v) Aqueous, Natural Poly Bottle, 500 mL

Representative image only.

Reagents Thymol Blue Indicator, 0.04% (w/v) Aqueous, Natural Poly Bottle, 500 mL

Reagents | Mfr: C7124500-500A| Item: UX-78914-47

Manufactured using the highest quality raw materials in ISO 9001 facility

$93.50USD / Each

$93.50USD / Each
Product Description

Key Features

  • Aqueous
  • Dark green liquid
  • Density: 1
  • Solubility information: miscible
  • 500 mL natural poly bottle
  • 2 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Two year warranty
Specifications
  • Concentration0.04% (w/v)
  • ClassIndicator
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)500
  • PackagingPoly bottle
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • Shelf Life (days)760
  • Shelf Life (months)24
  • Boiling Point C (° C)100
  • Boiling Point F (° F)212
  • Melting Point C (° C)0
  • Melting Point F (° F)32
  • DescriptionThymol Blue Indicator, 0.04% (w/v) Aqueous, Natural Poly Bottle, 500 mL
  • Warranty2 year
Documents & Certificates
Reviews
Q&A