Reagents Urea, Shotted Pellets, Natural Poly Container, 2.5 kg

Representative image only.

Reagents Urea, Shotted Pellets, Natural Poly Container, 2.5 kg

Reagents | Mfr: C2345500-2.5A5| Item: UX-78919-51

Manufactured using the highest quality raw materials in ISO 9001 facility

$140.75USD / Each

$140.75USD / Each
Product Description

Key Features

  • White shotted pellets
  • Density: 1.34
  • Boiling point: 135 °C (275 °F)
  • Melting point: 133 °C (271.4 °F), decomposes
  • Solubility information: 590 g/L at 20 °C (68 °F)
  • 2.5 kg natural poly container
  • 5 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

5 Year Warranty
Specifications
  • Linear FormulaNH2CONH2
  • ClassSalts
  • ChemistryOrganics
  • FormSolid
  • Weight (g)2500
  • Formula Weight60.06
  • PackagingPoly container
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number57-13-6
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • DescriptionUrea, Shotted Pellets, Natural Poly Container, 2.5 kg
  • Warranty5 years
Documents & Certificates
Reviews
Q&A