Reagents Zinc Chloride, Purified; 100 g

Representative image only.

Reagents Zinc Chloride, Purified; 100 g

Reagents | Mfr: C2352000-100C2| Item: UX-78914-67

Manufactured using the highest quality raw materials in ISO 9001 facility

$39.80USD / Each

$39.80USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs
  • 5 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

5 Year Warranty
Specifications
  • Purity GradePurified
  • Linear FormulaZnCl2
  • ClassSalts
  • FormSolid
  • Weight (g)100
  • Formula Weight136.29
  • Vapor Pressure (torr)0.9976
  • PackagingAmber Jar
  • DOT InformationZINC CHLORIDE, ANHYDROUS, 8
  • CAS Number7646-85-7
  • UN NumberUN2331
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • DescriptionZinc Chloride, 100g, Purified
  • Warranty5 years
Documents & Certificates
Reviews
Q&A