Retsch 01.462.0236 10 mL Mill Jar; stainless steel

Representative image only.

Retsch 01.462.0236 10 mL Mill Jar; stainless steel

Retsch | Mfr: 01.462.0236| Item: UX-04182-31

$1,940.00USD / Each

$1,940.00USD / Each

Buy It Together

Current Item

Retsch 01.462.0236 10 mL Mill Jar; stainless steel
Retsch 01.462.0236 10 mL Mill Jar; stainless steel
0418231

$1,940.00USD / Each
+ =
Total Price 1943.70 ADD 2 ITEMS TO CART
Product Description

More About this Item

Jars and media to meet your application needs.

Specifications
  • Sample Capacity (mL)10
  • MaterialStainless Steel
  • Description10 mL Mill Jar; stainless steel
Reviews
Q&A