Retsch 32.748.0005 Air-Jet Sieving Machine Industrial Vacuum Cleaner
Representative image only.
Retsch 32.748.0005 Air-Jet Sieving Machine Industrial Vacuum Cleaner
Retsch | Mfr: 32.748.0005| Item: UX-59996-37
Ideal for optimal use of your Air-Jet Sieving Machine
$2,780.00USD / Each
$2,780.00USD / Each
Buy It Together
Current Item
Retsch 32.748.0005 Air-Jet Sieving Machine Industrial Vacuum Cleaner
Retsch 32.748.0005 Air-Jet Sieving Machine Industrial Vacuum Cleaner
5999637
$2,780.00USD / Each
+
Retsch AS200 Jet Air-Jet Sieving Machine, 8" sieves
5999630
$15,230.00USD / Each
=
Total Price 18010.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Includes
manual vacuum regulator, sieve cover, rubber mallet, and EasySieve® software.
More About this Item
Sieving machine requires a vacuum cleaner to create the suction power, air-jet sieving machine, and 8" diameter test sieve, all sold separately. Other accessories are ideal for optimal perfomance of the sieving machine as well as maintenance. Air sieving machine is available as part number 59996-30.
Sieving machine requires a vacuum cleaner to create the suction power.
Specifications
  • DescriptionAir-Jet Sieving Machine Industrial Vacuum Cleaner