Ricca Chemical Company 8685-4 Trichloroacetic Acid, 10% (w/v) Aqueous Solution (120 mL)

Representative image only.

Ricca Chemical Company 8685-4 Trichloroacetic Acid, 10% (w/v) Aqueous Solution (120 mL)

Ricca Chemical Company | Mfr: 8685-4| Item: UX-86992-66

Quality and consistency your lab can count on

$43.30USD / Each

$43.30USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs
  • TRICHLOROACETIC ACID, 10%

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradeComponents all ACS
  • ClassGeneral Solutions
  • FormLiquid
  • Volume (mL)120
  • PackagingAmber Glass
  • DOT InformationTrichloroacetic Acid Solution; Class 8, Packing Group III
  • CAS NumberIncludes 76-0.-9 and 7732-18-5
  • UN NumberUN2564
  • Shelf Life (months)12
  • DescriptionTrichloroacetic Acid, 10% (w/v) Aqueous Solution (120 mL)
Reviews
Q&A