Shimpo DT-735 Portable, Light Weight Stroboscope with Rechargeable Battery
Representative image only.
Shimpo DT-735 Portable, Light Weight Stroboscope with Rechargeable Battery
Shimpo | Mfr: DT-735| Item: UX-98806-34
Rechargeable batteries allows unlimited measuring power
$625.00USD / Each
for an additional
$208.75USD / Each
for an additional
$293.50USD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
$625.00USD / Each
Buy It Together
Current Item
Shimpo DT-735 Portable, Light Weight Stroboscope with Rechargeable Battery
Shimpo DT-735 Portable, Light Weight Stroboscope with Rechargeable Battery
9880634
$625.00USD / Each
+
Shimpo SG-300 Strobe Gaurds
5356040
$103.50USD / Each
$115.00USD / Each
+
InnoCal NIST-Traceable Calibration; Stroboscope
1705010
$208.75USD / Each
=
Total Price 937.25 ADD 3 ITEMS TO CART
Product Description
Includes
one rechargeable NiMH battery and AC power cord.
Key Features
  • Lightweight aluminum construction allows for convenient one-handed operation
  • Repeatable accuaracy of ±0.01% of reading
  • Rechargeable internal batteries provide hours of operation
  • 1 year warranty
More About this Item
The portable stroboscopes are designed with a robust, extruded aluminum housing with a repeatable accuracy of ±0.01% of reading. Other features include a continuous duty cycle, microprocessor, external trigger, synchronous output, flash timer control, highly polished focused reflector, and manual flash controller knob.
The DT-735 heavy duty, battery powered stroboscope possesses the same great features of the DT-311D with the added convenience of total portability via its own internal power supply. The quick recharge battery lasts up to four hours of full continuous usage with its high intensity flash rate. The stroboscope provides power flexibility by utilizing either its rechargeable internal batteries or AC line-power when plugged into the included AC adapter/charger unit. These features make it ideal for quality inspection and preventive maintenance of automated motion equipment.
1 Year Warranty
Specifications
  • Min Flash Range (fpm)40
  • Max Flash Range (fpm)35000
  • Flash Accuracy (fpm)±0.01% of reading
  • Display Type5 Digit LED
  • Battery Type1 rechargeable NiMH (included)
  • DescriptionPortable, Light Weight Stroboscope with Rechargeable Battery