Shimpo FG-7FL Flat Adapter for FG Series Force Gauges
Representative image only.
Shimpo FG-7FL Flat Adapter for FG Series Force Gauges
Shimpo | Mfr: FG-7FL| Item: UX-93957-22
Choose from a range of accessories to assist you
$13.00USD / Each
$13.00USD / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
This replacement flat tip adapter comes standard with any FG-7000 series force gauge. It uses an M6 thread.
Please contact our Application Specialists for help in selecting an adapter tip for your testing needs.
1 Year Warranty
Specifications
  • DescriptionFlat Adapter for FG Series Force Gauges