Shimpo FG-7RD Extension Rod for FG Series Force Gauges
Representative image only.
Shimpo FG-7RD Extension Rod for FG Series Force Gauges
Shimpo | Mfr: FG-7RD| Item: UX-93957-26
Choose from a range of accessories to assist you
$19.00USD / Each
$19.00USD / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
This replacement tip extension rod comes standard with any FG-7000 series force gauge. It has M6 threads on each end.
Please contact our Application Specialists for help in selecting an adapter tip for your testing needs.
1 Year Warranty
Specifications
  • DescriptionExtension Rod for FG Series Force Gauges has M6 threads on each end.