Shimpo FG-7USB USB Cable for FG Series Force Gauges
Representative image only.
Shimpo FG-7USB USB Cable for FG Series Force Gauges
Shimpo | Mfr: FG-7USB| Item: UX-93957-20
Choose from a range of accessories to assist you
$13.00USD / Each
$13.00USD / Each
Product Description
Key Features
  • Ideal for for use with Shimpo FG-3000 and FG-7000 series Digital Force Gauges
  • 1 year warranty
More About this Item
This replacement item does not include the AC adapter (socket) required for connecting to a standard wall socket/power source. This is the USB cable only. The USB cable is used to charge the internal batteries, as well as connect the meter to a PC for data uploading. Simply connect the meter to the computer with the USB cable, then open the computer software.
Please contact our Application Specialists for help in selecting an adapter tip for your testing needs.
1 Year Warranty
Specifications
  • DescriptionUSB Cable for FG Series Force Gauges