Spex Acetic Acid Glacial, ACS Grade, Poly Bottle; 4 L

Representative image only.

Spex Acetic Acid Glacial, ACS Grade, Poly Bottle; 4 L

Spex | Item: UX-84410-02

Manufactured using the highest quality raw materials to ensure low-level impurities and consistent lot-to-lot quality

$201.00USD / Each

$201.00USD / Each
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Designed to work with any instrument and application
  • ACS grade—meets or exceeds purity standards set by the American Chemical Society
  • 4 L natural poly bottle

More About this Item

Flash point: 39°C
Density: 1.05
Molecular weight: 60.05 g/mol
Chemical formula: CH3COOH
Composition: glacial acetic acid 100%
Solubility: miscible
pH range: 2.4
Color: colorless liquid

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Hazardous shipping fee may apply.

Specifications
  • Purity GradeACS
  • Linear FormulaCH3COOH
  • ChemistrySolvents
  • Element/IonAcetic Acid
  • FormLiquid
  • SynonymsAcetic acid, Ethanoic acid, Methanecarboxylic acid, Vinegar acid
  • Volume (mL)4000
  • PackagingNatural poly bottle
  • DOT InformationHazardous; ACETIC ACID, GLACIAL; Class 8 (3), Packing Group II
  • Storage SymbolAmbient
  • Hazard SymbolFlammable, Corrosive
  • CAS Number64-19-7
  • UN NumberUN2789
  • Shelf Life (months)36
  • Boiling Point C (° C)117.9
  • Boiling Point F (° F)244.2
  • Melting Point C (° C)16.64
  • Melting Point F (° F)62
  • Refractive Index1.3577
  • DescriptionAcetic Acid Glacial, ACS Grade, Poly Bottle; 4 L
Documents & Certificates
Reviews
Q&A