Spex Hydrochloric Acid, 1 N, Poly Bottle; 1 L

Representative image only.

Spex Hydrochloric Acid, 1 N, Poly Bottle; 1 L

Spex | Item: UX-84410-31

Manufactured using the highest quality raw materials to ensure low-level impurities and consistent lot-to-lot quality

$38.00USD / Each

$38.00USD / Each
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Designed to work with any instrument and application
  • 1 L natural poly bottle

More About this Item

Solubility: miscible
Color: colorless liquid

Density: 1.02
Composition: water 96.30%, hydrochloric acid 3.70%
pH range: 0.1

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Hazardous shipping fee may apply.

Specifications
  • Concentration1 N
  • ChemistrySolvents
  • Element/IonHydrochloric Acid
  • FormLiquid
  • Volume (mL)1000
  • PackagingNatural poly bottle
  • DOT InformationHazardous; Hydrochloric Acid; Class 8, Packing Group III
  • Storage SymbolAmbient
  • Hazard SymbolCorrosive
  • CAS Number7647-01-0
  • UN NumberUN1789
  • Shelf Life (months)24
  • Boiling Point C (° C)100
  • Boiling Point F (° F)212
  • Melting Point C (° C)0
  • Melting Point F (° F)32
  • DescriptionHydrochloric Acid, 1 N, Poly Bottle; 1 L
Documents & Certificates
Reviews
Q&A