Spex ICAL-2 Instrument Calibration Standard 2 in 5% HNO3; 125 mL

Representative image only.

Spex ICAL-2 Instrument Calibration Standard 2 in 5% HNO3; 125 mL

Spex | Item: UX-04568-96

Multi-element standards for ICP and ICP-MS instrumentation

$217.00USD / Each

$217.00USD / Each
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025/2017 and ISO 17034:2016
  • Certified to DQS to ISO 9001:2015
  • Calibrate ICP and ICP-MS instrumentation prior to analysis
  • One calibration standard must be at the Contract Required Quantification Limit (CRQL) for ISM 01.2
  • Multi-element instrument calibration standard 2 for ICP & ICP-MS in 5% HNO3 matrix
  • 125 mL bottle

More About this Item

Our Contract Laboratory Program (CLP) standards allow you to Calibrate with Confidence®. The standards are to be used in conjunction with the Statement of Work for Inorganic Analysis; Multi-Media/Multi-Concentration Document Number ILM 05.3/ISM 01.2.

US EPA SOW ILM 05.3/ISM 01.2 gives specific procedures for the methods of analysis, target elements, and concentration levels. Standards are specified not only by the elements present and their relative concentrations, but also the order and frequency of running standards, blanks and samples. Details of these specifications may be found in the US EPA SOW ILM 05.3/ISM 01.2 in the following sections:
• Exhibit C, Inorganic Target Analyte List (TAL)
• Exhibit D, Analytical Methods
• Exhibit E, QA/QC Requirements

Solutions may be used separately or mixed together for preparation of the analytical curve. When mixed, these solutions will yield a standard containing all of the elements in the Target Analyte List (TAL). Instruments must be calibrated daily, every 24 hours, or each time the instrument is set-up. Calibration standards must be prepared fresh for each analysis and discarded after use. A dilution of 100-fold is suggested. Antimony and mercury can be diluted as required.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

This multi-element standard contains 5 elements.

• Instrument Calibration Standard 2 is a 125 mL bottle which contains 400 µg/mL (400 ppm) Nickel; 200 µg/mL (200 ppm) Zinc; 150 µg/mL (150 ppm) Manganese; and 100 µg/mL (100 ppm) Chromium, Silver.

Hazardous material shipping fees may apply.

Specifications
  • ConcentrationMultiple
  • Matrix5% HNO3
  • Element/IonMultiple
  • FormLiquid
  • Volume (mL)125
  • DOT InformationHazardous; Corrosive liquid, acidic, inorganic, n.o.s, (Nitric acid solution); Class 8, Packing Group III
  • Storage SymbolAmbient
  • Hazard SymbolCorrosion
  • UN NumberUN3264
  • Shelf Life (months)12
  • DescriptionInstrument Calibration Standard 2 in 5% HNO3; 125 mL
Documents & Certificates
Reviews
Q&A