Spex Isopropanol (2-Propanol; Isopropyl Alcohol), ACS Grade; 4 L

Representative image only.

Spex Isopropanol (2-Propanol; Isopropyl Alcohol), ACS Grade; 4 L

Spex | Item: UX-84410-37

Manufactured using the highest quality raw materials to ensure low-level impurities and consistent lot-to-lot quality

$206.00USD / Each

$206.00USD / Each
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Designed to work with any instrument and application
  • ACS grade—meets or exceeds purity standards set by the American Chemical Society
  • 4 L amber glass bottle

More About this Item

Flash point: 12 °C
Density: 0.78
Molecular weight: 60.1 g/mol
Solubility: miscible
Refractive index: 1.376 to 1.378nD
Color: colorless liquid

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Hazardous shipping fee may apply.

Specifications
  • Purity GradeACS
  • ChemistrySolvents
  • Element/IonIsopropanol
  • FormLiquid
  • Volume (mL)4000
  • PackagingAmber glass bottle
  • DOT InformationHazardous; Isopropanol; Class 3, Packing Group II
  • Storage SymbolAmbient
  • Hazard SymbolFlammable, Irritant
  • CAS Number67-63-0
  • UN NumberUN1219
  • Shelf Life (months)60
  • Boiling Point C (° C)82.5
  • Boiling Point F (° F)180.5
  • Melting Point C (° C)-90
  • Melting Point F (° F)-130
  • DescriptionIsopropanol (2-Propanol; Isopropyl Alcohol), ACS Grade; 4 L
Documents & Certificates
Reviews
Q&A