Spex MPK-AA-5PK Acetic Acid Mobile Phase Modifier for HPLC/LCMS, Neat; 5 x 1 mL

Representative image only.

Spex MPK-AA-5PK Acetic Acid Mobile Phase Modifier for HPLC/LCMS, Neat; 5 x 1 mL

Spex | Item: UX-41018-02

Mobile phase modifiers for HPLC and LC/MS

$56.00USD / Pkg of 5

$56.00USD / Pkg of 5
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Conveniently packaged for error-free use
  • Designed to be directly added to mobile phase reservoirs for easy use
  • Ability to mix and match modifiers to create personalized method development kits

More About this Item

High Performance Liquid Chromatography (HPLC) is one of the most common instrumental techniques for analyzing mixtures of organic compounds in analytical labs around the world. HPLC is a complex process that depends on each molecule’s ability to interact with the mobile phase that carries it through the column, the solid phase that fills the columns, as well as interactions between the different molecules in the sample.

These interactions are particularly important in LCMS when the molecules being analyzed need ionizable functionality. Many labs are now using reverse-phase high performance chromatography (RP-HPLC), where the stationary phase is hydrophobic or nonpolar. However, reverse-phase columns may contain open silanol groups, which may interact with polar molecules. This leads to reduced resolution, tailing of peaks and reduction in column life.

The addition of modifiers, like trifluoracetic, acetic and formic acid, and the corresponding ammonium salts, are widely used to improve the quality of RP-HPLC chromatography analyses. These modifiers adjust the pH of the mobile phase and provide the chemist with the ability to control the level of ionization of each molecule.

Modifiers can form ion pairs with the Lewis-basic and Lewis-acidic sites on the solvent-accessible regions of the stationary phase. This decreases the opportunity for analytes to interact with these regions and results in increased column longevity and performance.

Spex mobile phase modifiers are conveniently packaged for error-free use. Our ampules contain precisely measured aliquots of high purity modifiers which are designed to be directly added to your mobile phase for easy use. Customers are encouraged to mix and match modifiers to create personalized method development kits.
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Hazardous material shipping fees may apply.

This item needs to be refrigerated immediately upon arrival.

Specifications
  • MatrixNeat
  • Element/IonAcetic Acid
  • Volume (mL)1
  • Packaging5 x 1 mL
  • Storage SymbolRefrigerator
  • CAS Number64-19-7
  • Shelf Life (days)365
  • Shelf Life (months)12
  • DescriptionAcetic Acid Mobile Phase Modifier for HPLC/LCMS, Neat; 5 x 1 mL
Documents & Certificates
Reviews
Q&A