Spex PLBLK-H2O-2L Assurance® Grade DI Water Blank for AA and ICP in H2O, 2 L
Representative image only.
Spex PLBLK-H2O-2L Assurance® Grade DI Water Blank for AA and ICP in H2O, 2 L
Spex | Item: UX-04567-50
Establish your baseline for AA and ICP analysis
$146.00USD / Each
$146.00USD / Each
Product Description
Includes
Supplier Certificate of Analysis (CoA) is included with product shipment.
Key Features
  • Can be run directly as a blank to establish your baseline
  • May be used to dilute your multi-element standards
  • DI water blank
More About this Item
Assurance® grade certified calibration and matrix blanks are designed for atomic absorption (AA) and inductively coupled plasma (ICP) analysis. Blanks available include DI water, hydrochloric acid, or nitric acid.
Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.
Do not use any acid or water as a diluent if you are not certain of its purity.
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
Specifications
  • Purity GradeAssurance® Grade
  • MatrixH2O
  • ClassInorganic Standards
  • Volume (mL)2
  • DescriptionDI Water Blank for AA and ICP in H2O, 2 L