Spex PLBLK-HCL Assurance® Grade Hydrochloric Acid Blank for AA and ICP in 5% HCl, 500 mL

Representative image only.

Spex PLBLK-HCL Assurance® Grade Hydrochloric Acid Blank for AA and ICP in 5% HCl, 500 mL

Spex | Item: UX-04567-47

Establish your baseline for AA and ICP analysis

$89.00USD / Each

$89.00USD / Each
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Can be run directly as a blank to establish your baseline
  • May be used to dilute your multi-element standards
  • Hydrochloric acid blank

More About this Item

Assurance® grade certified calibration and matrix blanks are designed for atomic absorption (AA) and inductively coupled plasma (ICP) analysis. Blanks available include DI water, hydrochloric acid, or nitric acid.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Hazardous material shipping fees may apply.

Do not use any acid or water as a diluent if you are not certain of its purity.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradeAssurance® Grade
  • Matrix5% HCl
  • ClassInorganic Standards
  • Volume (mL)500
  • DOT InformationHazmat Fees May Apply
  • DescriptionHydrochloric Acid Blank for AA and ICP in 5% HCl, 500 mL
Reviews
Q&A