Spex PLBLK-HNO3 Assurance® Grade Nitric Acid Blank for AA and ICP in 5% HNO3, 500 mL

Representative image only.

Spex PLBLK-HNO3 Assurance® Grade Nitric Acid Blank for AA and ICP in 5% HNO3, 500 mL

Spex | Item: UX-04567-46

Establish your baseline for AA and ICP analysis

$100.00USD / Each

$100.00USD / Each
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Can be run directly as a blank to establish your baseline
  • May be used to dilute your multi-element standards
  • Nitric acid blank

More About this Item

Assurance® grade certified calibration and matrix blanks are designed for atomic absorption (AA) and inductively coupled plasma (ICP) analysis. Blanks available include DI water, hydrochloric acid, or nitric acid.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Hazardous material shipping fees may apply.

Do not use any acid or water as a diluent if you are not certain of its purity.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Causes severe skin burns and eye damage. Causes serious eye damage. Do not breathe dusts or mists. If on skin (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. If in eyes: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER/doctor. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations.

Corrosive
Specifications
  • Purity GradeAssurance® Grade
  • Matrix5% HNO3
  • ClassInorganic Standards
  • Volume (mL)500
  • DOT InformationHazmat Fees May Apply
  • DescriptionNitric Acid Blank for AA and ICP in 5% HNO3, 500 mL
Reviews
Q&A