Spex Potassium Permanganate, 0.1 N; 4 L

Representative image only.

Spex Potassium Permanganate, 0.1 N; 4 L

Spex | Item: UX-84410-65

Manufactured using the highest quality raw materials to ensure low-level impurities and consistent lot-to-lot quality

$100.00USD / Each

$100.00USD / Each
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Designed to work with any instrument and application
  • 4 L amber glass bottle

More About this Item

Density: 1
Composition: water 99.68%, potassium permanganate 0.32%
Solubility: miscible
Color: purple liquid

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Concentration0.1 N
  • ChemistrySolvents
  • Element/IonPotassium Permanganate
  • FormLiquid
  • Volume (mL)4000
  • PackagingAmber glass bottle
  • DOT InformationNonhazardous; Not Regulated
  • Storage SymbolAmbient
  • Hazard SymbolNot Regulated
  • CAS Number7722-64-7
  • UN NumberNot Regulated
  • Shelf Life (months)18
  • Boiling Point C (° C)100
  • Boiling Point F (° F)212
  • Melting Point C (° C)32
  • Melting Point F (° F)0
  • DescriptionPotassium Permanganate, 0.1 N; 4 L
Documents & Certificates
Reviews
Q&A