Spex Silver Nitrate, 0.1 N, Amber Glass Bottle; 4 L

Representative image only.

Spex Silver Nitrate, 0.1 N, Amber Glass Bottle; 4 L

Spex | Item: UX-84410-69

Manufactured using the highest quality raw materials to ensure low-level impurities and consistent lot-to-lot quality

$501.00USD / Each

$501.00USD / Each
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Designed to work with any instrument and application
  • 0.1 N silver nitrate in 4 L amber glass bottle

More About this Item

Solubility: miscible
Color: colorless liquid

Density: 1.01
Composition: water 98.32%, silver nitrate 1.68%

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Concentration0.1 N
  • ChemistrySolvents
  • Element/IonSilver Nitrate
  • FormLiquid
  • Volume (mL)4000
  • PackagingAmber glass bottle
  • DOT InformationNonhazardous; Not Regulated
  • Storage SymbolAmbient
  • Hazard SymbolEnvironment
  • UN NumberNot Regulated
  • CAS Number7761-88-8
  • Shelf Life (months)24
  • Boiling Point C (° C)100
  • Boiling Point F (° F)212
  • Melting Point C (° C)0
  • Melting Point F (° F)32
  • DescriptionSilver Nitrate, 0.1 N, Amber Glass Bottle; 4 L
Documents & Certificates
Reviews
Q&A