Spex Sodium Hydroxide, 0.1 N, Cubitainer® Container; 20 L

Representative image only.

Spex Sodium Hydroxide, 0.1 N, Cubitainer® Container; 20 L

Spex | Item: UX-84410-77

Manufactured using the highest quality raw materials to ensure low-level impurities and consistent lot-to-lot quality

$223.00USD / Each

$223.00USD / Each
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Designed to work with any instrument and application
  • 0.1 N sodium hydroxide in 20 L Cubitainer® container

More About this Item

Solubility: miscible
Color: colorless liquid

Density: 1
Composition: water 99.60%, sodium hydroxide 0.40%
pH Range: 13

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Concentration0.1 N
  • ChemistrySolvents
  • Element/IonSodium Hydroxide
  • FormLiquid
  • Volume (mL)20000
  • PackagingCubitainer® container
  • DOT InformationNonhazardous; Not Regulated
  • Storage SymbolAmbient
  • Hazard SymbolNot Regulated
  • UN NumberNot Regulated
  • CAS Number1310-73-2
  • Shelf Life (months)24
  • Boiling Point C (° C)100
  • Boiling Point F (° F)212
  • Melting Point C (° C)0
  • Melting Point F (° F)32
  • DescriptionSodium Hydroxide, 0.1 N, Cubitainer® Container; 20 L
Documents & Certificates
Reviews
Q&A