Sussman 39123C Replacment Heating Element, 12 kW, 240 VAC

Representative image only.

Sussman 39123C Replacment Heating Element, 12 kW, 240 VAC

Sussman | Mfr: 39123C| Item: UX-07602-31

$931.00USD / Each

$931.00USD / Each
Product Description

Key Features

  • 1 year warranty

More About this Item

Keep your steam generator running smoothly with these parts and accessories.

1 Year Warranty
Specifications
  • Compatible WithSussman Steam Generator with 12 kW, 240 VAC power
  • Power (VAC)240
  • DescriptionReplacment Heating Element, 12 kW, 240 VAC
  • Warranty1 year
Reviews
Q&A