TEL UK Air Flow Monitor for Fume Hoods, 30 to 400 FPM Monitoring Range
Representative image only.
TEL UK Air Flow Monitor for Fume Hoods, 30 to 400 FPM Monitoring Range
TEL UK | Mfr: M52TEL53US| Item: UX-33556-50
Reliable, durable sensor element
$706.00USD / Each
$706.00USD / Each
Product Description
Includes
low voltage transformer and mounting bracket for surface mount. Call our Application Specialists if a flush mount requiring a 2" x 3" (5.1 x 7.6 cm) opening is preferred.
Key Features
  • Simple field calibration
  • Multiple alarms—audible, visual, and relay
More About this Item
Monitors maintain accurate response even when using low-flow hood conditions. Alarms activate based on sustained low-flow condition. Use pushbuttons to set alarm and perform easy two-point calibration. Monitors will also indicate high sash condition (requires proximity switch, not included) with configurable time delay. Record hood performance using RS-232 connection to computer.
Model 33556-50 features a horn alarm with green and red LED indicators. The graphic display model 33556-60 comes with an external velocity meter and shows current velocity in fpm with bar graph depiction of recent hood airflow performance; calibration instructions appear on screen. Choose model 33556-70 for its extra wide measurement range. It includes a remote velocity probe which can be installed in your ductwork for use with biosafety cabinets.
Specifications
  • Power (VAC)120
  • Power (Hz)60
  • DescriptionAir Flow Monitor for Fume Hoods, 30 to 400 FPM Monitoring Range