Thermo Scientific Barnstead 09.4000 Double Cartridge Pretreatment System
Representative image only.
Thermo Scientific Barnstead Mfr # 09.4000 – Item # EW-99299-38
Multiple outputs available to meet your application needs
  • For Use With: LabTower TII, Pacific RO, and Pacific TII
  • 3 month warranty
PRODUCT OPTIONS
$950.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Description3 holder pretreatment package
  • Warranty3 month
3 Month Warranty
Includes
5µm filter with carbon cartridge and a hardness stabilizer cartridge.
Key Features
  • For Use With: LabTower TII, Pacific RO, and Pacific TII
  • 3 month warranty
More About this Item
Sterile vents and pretreatment cartridges round out your purification system.
Double cartridge pretreatment system (includes 5µm filter with carbon cartridge and a hardness stabilizer cartridge).