Thermo Scientific CODHP0 High range COD kit, 0 to 15,000 ppm
Representative image only.
Thermo Scientific CODHP0 High range COD kit, 0 to 15,000 ppm
Thermo Scientific | Mfr: CODHP0| Item: UX-99581-44
Optimize water analysis
$70.20USD / Each
$70.20USD / Each
Product Description
Key Features
  • Choose from a variety of accessories
More About this Item
Get the most out of your water analysis equipment with these quality solutions and accessories.
Use for EPA reporting of COD test results and general monitoring. The pre-measured reagents offer a long shelf life. Each kit contains 25 vials.
EPA United States Environmental protection agency
Specifications
  • Compatible WithThermo Scientific Orion COD Reactor & Colorimeter
  • DescriptionHigh range COD kit, 0 to 15,000 ppm