Thermo Scientific GENESYS™ 8-Position Carousel, 10 mm, for GENESYS™ 100 Series and BioMate 160
Representative image only.
Thermo Scientific GENESYS™ 8-Position Carousel, 10 mm, for GENESYS™ 100 Series and BioMate 160
Thermo Scientific | Mfr: 840-303400| Item: UX-02654-37
Enhance the versitility of your spectrophotometer
$445.00USD / Each
$445.00USD / Each
Buy It Together
Current Item
Thermo Scientific GENESYS™ 8-Position Carousel, 10 mm, for GENESYS™ 100 Series and BioMate 160
Thermo Scientific GENESYS™ 8-Position Carousel, 10 mm, for GENESYS™ 100 Series and BioMate 160
0265437
$445.00USD / Each
+ =
Total Price 4535.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Expand your spectrophotometer capacity and capabilities
  • 2 year warranty
More About this Item
Choose from a wide range of replacement parts and accessories including cuvette holders, sipper pumps, and more suitable for use with the Thermo Scientific™ GENESYS™ visible and UV-visible spectrophotometers. Spectrophotometers sold separately.
Increase throughput in busy labs with this 8-position cell carousel. The magnetic latch allows for easy echange. This cell holder is compatible with 10 mm path length cells only — with 4.5L cell and semi-micro cells. The cell windows must have a ≥3 mm width and ≥1000 µL volume.
Available for sale and use in the United States only.
Two year warranty
Specifications
  • Compatible WithGENESYS 140, 150, 180, and 160 BioMate spectrophotometers
  • Description8-Position Carousel, 10 mm, for GENESYS™ 100 Series and BioMate 160