Thermo Scientific GENESYS™ Long Path Rectangular Cell Holder for GENESYS™ 100 Series and BioMate 160
Representative image only.
Thermo Scientific GENESYS™ Long Path Rectangular Cell Holder for GENESYS™ 100 Series and BioMate 160
Thermo Scientific | Mfr: 840-303800| Item: UX-02654-39
Enhance the versitility of your spectrophotometer
$284.50USD / Each
$284.50USD / Each
Buy It Together
Current Item
Thermo Scientific GENESYS™ Long Path Rectangular Cell Holder for GENESYS™ 100 Series and BioMate 160
Thermo Scientific GENESYS™ Long Path Rectangular Cell Holder for GENESYS™ 100 Series and BioMate 160
0265439
$284.50USD / Each
+ =
Total Price 4374.50 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Expand your spectrophotometer capacity and capabilities
  • 2 year warranty
More About this Item
Choose from a wide range of replacement parts and accessories including cuvette holders, sipper pumps, and more suitable for use with the Thermo Scientific™ GENESYS™ visible and UV-visible spectrophotometers. Spectrophotometers sold separately.
Long path rectangular cell holder is suitable for 10 to 100 mm pathlength cuvettes.
Available for sale and use in the United States only.
Two year warranty
Specifications
  • Compatible WithGENESYS 140, 150, 180, and 160 BioMate spectrophotometers
  • DescriptionLong Path Rectangular Cell Holder for GENESYS™ 100 Series and BioMate 160