Thermo Scientific R111101 Prepared Plates, TSA w/Lecithin/Polysorbate Agar; 20/Pk
Representative image only.
Thermo Scientific R111101 Prepared Plates, TSA w/Lecithin/Polysorbate Agar; 20/Pk
Thermo Scientific | Mfr: R111101| Item: UX-14207-66
Stringent quality control ensures high-quality performance as intended!
$246.50USD / Pkg of 20
$246.50USD / Pkg of 20
Product Description
Key Features
  • Available for sale in the United States only
  • For lab use only; not intended for in vitro purposes
  • Made using GripFit™ technology, which ensures the lid remains in place during handling and yet can be easily removed during environmental monitoring
More About this Item
Choose from a variety of media formulations. Use this culture media for the isolation and differentiation of microorganisms from samples or specimens. Each raw component is pretested before being used, for quality assurance and consistency from lot batch. Intended for lab practice only; not intended for in vitro purposes. Plates have stacking rings which interlock to minimize sliding.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
Specifications
  • Media Name (No.)Tryptic Soy w/ Lecithin, Polysorbate 80
  • DescriptionPrepared Plates, TSA w/Lecithin/Polysorbate Agar; 20/Pk