Trajan Premium High-Temp Center Guide GC Septa, 17 mm OD; 50/PK
Representative image only.
Trajan Premium High-Temp Center Guide GC Septa, 17 mm OD; 50/PK
Trajan | Mfr: 298817| Item: UX-01994-46
Optimized for the most demanding GC and GC/MS applications
$122.25USD / Pkg of 50
$122.25USD / Pkg of 50
Product Description
Key Features
  • Formulated to extend low-bleed and outstanding mechanical properties
More About this Item
(Bleed and Temperature Optimized) are designed for low bleed and temperatures up to 400°C (752 °F). Septa changes are easy—septa will not stick to your port.