Trajan SGE Replacement Plunger, Compatible with PerkinElmer Fixed Needle Gas Tight Syringe,1 mL
Representative image only.
Trajan SGE Replacement Plunger, Compatible with PerkinElmer Fixed Needle Gas Tight Syringe,1 mL
Trajan | Mfr: 0318441| Item: EW-16002-64
Choose wisely to save your operational cost—no need to buy new syringes instead
$94.10USD / Pkg of 1
$94.10USD / Pkg of 1
Buy It Together
Current Item
Trajan SGE Replacement Plunger, Compatible with PerkinElmer Fixed Needle Gas Tight Syringe,1 mL
Trajan SGE Replacement Plunger, Compatible with PerkinElmer Fixed Needle Gas Tight Syringe,1 mL
1600264
$94.10USD / Pkg of 1
+ =
Total Price 226.60 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Each stainless steel plunger is individually fitted to each barrel
  • 6 month warranty
More About this Item
All SGE gas-tight syringes have replaceable plungers.
The standard plunger in MicroVolume syringes is not replaceable.
6 Month Warranty
Specifications
  • DescriptionTrajan SGE Replacement GT plunger and 1/4-28 UNF Termination for PerkinElmer 1 mL Syringe; 1/Ea