US Industrial SDT7200/12V/D Hydrogen Transmitter, 0 to 20,000 ppm, 4 to 20 mA output

Representative image only.

US Industrial SDT7200/12V/D Hydrogen Transmitter, 0 to 20,000 ppm, 4 to 20 mA output

US Industrial | Mfr: SDT7200/12V/D| Item: UX-10150-43

Provides a broad range for toxic (PPM) and explosive gases (%LEL) detector

$1,430.00USD / Each

$1,430.00USD / Each
Product Description

Includes

sensor and controller (3" diameter) in the explosion-proof enclosure for use in hazardous location (Class 1, Division 1, Groups B, C, and D)

Key Features

  • Explosion-proof housing with see-through window meets NEC requirements for Class 1, Division 1, Groups B, C, and D
  • 1 year warranty

More About this Item

The SDT transmitter is designed for stand alone monitoring of hazardous gases and vapors. The gas level is indicated on the 4-digit, red LED display and transmitted with the linear 4-20 mA output for data loggers or recorders. Semiconductor sensors measure toxic gases in the low ppm ranges. They feature long sensor life and fast response time. Sensors are not affected by temperature and humidity changes — they can be used both indoors and outdoors. Catalytic sensors monitor flammable gases in the %LEL, or volume percent range. These sensors feature fast response, high accuracy, long life, and a high resistance to poisoning.

1 Year Warranty
Specifications
  • Gases MeasuredHydrogen
  • Accuracy±5% of measuring range
  • Output4 to 20 mA
  • Display Type4-digits, red LED
  • Power (VDC)12
  • Length (in)4
  • Width (in)4 1/2
  • DescriptionHydrogen Transmitter, 0 to 20,000 ppm, 4 to 20 mA output
  • Warranty1 year
Reviews
Q&A