Vector Laboratories SK-5100 Red Substrate Kit, Magenta, 200 ml

Representative image only.

Vector Laboratories SK-5100 Red Substrate Kit, Magenta, 200 ml

Vector Laboratories | Mfr: SK-5100| Item: UX-93952-02

Successfully label multiple antigens in same tissue section

$355.00USD / Each

$355.00USD / Each
Product Description

Key Features

  • Dropper bottles simplify handling of chromogens
  • No wait times for mixing and dissolving powders or tablets
  • 1 year warranty

More About this Item

These enzyme substrate kits for use with alkaline phosphatase detection systems offer a wide variety of applications. Numerous color choices are available for staining antigens in pigmented tissues and melanomas, as well as for localizing multiple antigens in the same tissue section. Viewing options for the substrates include brightfield, fluorescence, darkfield, electron microscopy, and spectral imaging.

Vector Red and ImmPACT Red Substrate Kits produce red, heat stable reaction products that are highly fluorescent and non-fading. Because of their stability under ISH conditions, both chromogens can be used in IHC/ISH double labeling applications.

1 Year Warranty
Specifications
  • ColorMagenta
  • Volume (mL)200
  • DescriptionRed Substrate Kit, Magenta, 200 ml
  • Warranty1 year
Reviews
Q&A