Velp A00001306 SS Stirring Shaft with Fixed Blade
Representative image only.
Velp A00001306 SS Stirring Shaft with Fixed Blade
Velp | Mfr: A00001306| Item: UX-50200-82
$157.50USD / Each
$157.50USD / Each
Buy It Together
Current Item
Velp A00001306 SS Stirring Shaft with Fixed Blade
Velp A00001306 SS Stirring Shaft with Fixed Blade
5020082
$157.50USD / Each
+ + =
Total Price 1952.50 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • 2 year warranty
More About this Item
Choose from our comprehensive line of Velp mixer accessories including stirring shafts, support base, and clamps.
Blade generates axial flow in container from top towards the bottom. Recommended for use at medium-high speeds for spinning light solids, for flocculation, mixing thickening agents, sludge stirring, etc.
Two year warrantyConformite Europeenne
Specifications
  • DescriptionSS Stirring Shaft with Fixed Blade