Velp A00001309 SS Stirring Shaft with Turbine
Representative image only.
Velp A00001309 SS Stirring Shaft with Turbine
Velp | Mfr: A00001309| Item: UX-50200-85
$285.00USD / Each
$285.00USD / Each
Buy It Together
Current Item
Velp A00001309 SS Stirring Shaft with Turbine
Velp A00001309 SS Stirring Shaft with Turbine
5020085
$285.00USD / Each
+ + =
Total Price 2080.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • 2 year warranty
More About this Item
Choose from our comprehensive line of Velp mixer accessories including stirring shafts, support base, and clamps.
Stirring shaft generates a radial flow which leads to suction of the product from top towards the bottom with high turbulence and high shearing forces.
Two year warrantyConformite Europeenne
Specifications
  • DescriptionSS Stirring Shaft with Turbine