Velp A00001311 Mixer Tangential Flow Stirring Shaft, SS, Anchor-Style
Representative image only.
Velp A00001311 Mixer Tangential Flow Stirring Shaft, SS, Anchor-Style
Velp | Mfr: A00001311| Item: UX-50200-87
$141.00USD / Each
$141.00USD / Each
Buy It Together
Current Item
Velp A00001311 Mixer Tangential Flow Stirring Shaft, SS, Anchor-Style
Velp A00001311 Mixer Tangential Flow Stirring Shaft, SS, Anchor-Style
5020087
$141.00USD / Each
+ + =
Total Price 1936.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • 2 year warranty
More About this Item
Choose from our comprehensive line of Velp mixer accessories including stirring shafts, support base, and clamps.
Stirring shaft generates a tangential flow with high shearing forces with minimal sedimentation on container walls.
Two year warrantyConformite Europeenne
Specifications
  • Material316 Stainless Steel
  • Impeller Size (in)1.77
  • Impeller Size (mm)45
  • DescriptionMixer Tangential Flow Stirring Shaft, SS, Anchor-Style