Whatman 6700-7502 Polycap TF PTFE Capsule Filter, 0.2um, 1000cm2, 1/4-3/8" ID; 1/Pk
Representative image only.
Whatman 6700-7502 Polycap TF PTFE Capsule Filter, 0.2um, 1000cm2, 1/4-3/8" ID; 1/Pk
Whatman | Mfr: 6700-7502| Item: UX-29700-12
PTFE membrane is ideal for venting, in-line filtration, and isolation (gas pass, liquids, and aerosol stopped)
$213.95USD / Each
$213.95USD / Each
Product Description
Key Features
  • Autoclavable
More About this Item
Capsules feature sturdy PP housings with fused seals—no glues or adhesives to contaminate filtrate. Capsules include a luer lock vent to guarantee that no filtration area is wasted. Use vent to draw solution into the capsule or let air escape from the capsule or sample port.
CYTIVA Whatman™ Warranties to served market customers are in lieu of all other warranties, expressed or implied, and CYTIVA Whatman™ disclaims all other warranties, including without limitation, the implied warranties of non-infringement, merchantability, fitness for a particular purpose, quiet enjoyment, system integration and data accuracy. CYTIVA Whatman™ limited warranties set forth in the CYTIVA Whatman™ warranty statement shall be enforced by and only by the served market customers.
Specifications
  • Membrane MaterialPTFE
  • Pore Size (µm)0.2
  • Effective Filter Area (cm²)1000
  • Connections1/4" to 3/8" stepped barb
  • DescriptionPolycap TF PTFE Capsule Filter, 0.2um, 1000cm2, 1/4-3/8" ID; 1/Pk